5-Phenyl[1,2,4]oxadiazole-3-carboxylic acid ethyl ester 95%
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 5-phenyl-1,2,4-oxadiazole-3-carboxylate
OR300703-1G
OR300703-5G
APOSOR300703-1GEA
5560
CZK
APOSOR300703-1G
APOSOR300703-5G
5-Phenyl[1,2,4]oxadiazole-3-carboxylic acid ethyl ester 95%
5-Phenyl[1,2,4]oxadiazole-3-carboxylic acid ethyl ester
Vzorec:
C₁₁H₁₀N₂O₃ MW: 218,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01764002 CAS číslo: 37384-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...