Tisk…

2-Bromo-4'-(methylsulphonyl)acetophenone

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Bromo-1-(4-(methylsulphonyl)phenyl)ethanone, 4-(Methylsulphonyl)phenacyl bromide

OR30069-1G OR30069-25G OR30069-5G
APOSOR30069-1GEA 622 CZK
APOSOR30069-1G APOSOR30069-25G APOSOR30069-5G
2-Bromo-4'-(methylsulphonyl)acetophenone
2-Bromo-4'-(methylsulphonyl)acetophenone
Vzorec: C₉H₉BrO₃S
MW: 277,1350 g/mol
Bod tání: 125…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00673134
CAS číslo: 50413-24-6
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR