Ethyl-3-chloromethyl[1,2,4]oxadiazole-5-carboxylate 95%
Dodavatel: Apollo Scientific
OR300686-10G
OR300686-1G
OR300686-5G
APOSOR300686-10GEA
28110
CZK
APOSOR300686-10G
APOSOR300686-1G
APOSOR300686-5G
Ethyl-3-chloromethyl[1,2,4]oxadiazole-5-carboxylate 95%
Ethyl-3-chloromethyl[1,2,4]oxadiazole-5-carboxylate
Vzorec:
C₆H₇ClN₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD09749852 CAS číslo: 25977-19-9 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...