Tisk…

2-Aminobenzothiazole-6-carbonitrile

Dodavatel: Apollo Scientific
Varování

OR300670-1G OR300670-5G
APOSOR300670-1GEA 2440 CZK
APOSOR300670-1G APOSOR300670-5G
2-Aminobenzothiazole-6-carbonitrile
2-Aminobenzothiazole-6-carbonitrile
Vzorec: C₈H₅N₃S
MW: 175,2104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664359
CAS číslo: 19759-66-1
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR