Tisk…

6-Nitro-o-anisidine

Dodavatel: Apollo Scientific

OR300658-1G OR300658-5G
APOSOR300658-1GEA 622 CZK
APOSOR300658-1G APOSOR300658-5G
6-Nitro-o-anisidine
6-Nitro-o-anisidine
Vzorec: C₇H₈N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD01930197
CAS číslo: 16554-45-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR