Tisk…

Morpholin-4-yl(phenyl)acetonitrile

Dodavatel: Apollo Scientific

OR300647-250MG OR300647-5G OR300647-10G OR300647-25G
APOSOR300647-250MGEA 1660 CZK
APOSOR300647-250MG APOSOR300647-5G APOSOR300647-10G APOSOR300647-25G
Morpholin-4-yl(phenyl)acetonitrile
Morpholin-4-yl(phenyl)acetonitrile
Vzorec: C₁₂H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD00014619
CAS číslo: 15190-10-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR