Tisk…

Isoxazol-5-ylamine

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Aminoisoxazole, Isoxazol-5-amine

OR300646-1G OR300646-250MG OR300646-5G
APOSOR300646-1GEA 1580 CZK
APOSOR300646-1G APOSOR300646-250MG APOSOR300646-5G
Isoxazol-5-ylamine
Isoxazol-5-ylamine
Vzorec: C₃H₄N₂O
MW: 84,08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00128192
CAS číslo: 14678-05-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR