Tisk…

7-Bromobenzo[b]thiophene

Dodavatel: Apollo Scientific
Varování

OR300641-1G OR300641-5G
APOSOR300641-1GEA 0 CZK
APOSOR300641-1G APOSOR300641-5G
7-Bromobenzo[b]thiophene
7-Bromobenzo[b]thiophene
Vzorec: C₈H₅BrS
Storage Temperature: Refrigerator
MDL Number: MFCD06657729
CAS číslo: 1423-61-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR