3-Chloromethyl-5-methyl[1,2,4]oxadiazole 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3-(Chloromethyl)-5-methyl-1,2,4-oxadiazole
OR300629-1G
APOSOR300629-1GEA
5460
CZK
APOSOR300629-1G
3-Chloromethyl-5-methyl[1,2,4]oxadiazole 98%
3-Chloromethyl-5-methyl[1,2,4]oxadiazole
Vzorec:
C₄H₅ClN₂O MW: 132,55 g/mol Bod varu: 42…43 °C/0,5mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD01809083 CAS číslo: 1192-80-9 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...