Tisk…

Methyl-3-(benzylamino)propanoate

Dodavatel: Apollo Scientific
Varování

OR300523-10G OR300523-1G OR300523-5G OR300523-25G OR300523-250MG OR300523-2.5G
APOSOR300523-10GEA 2610 CZK
APOSOR300523-10G APOSOR300523-1G APOSOR300523-5G APOSOR300523-25G APOSOR300523-250MG APOSOR300523-2.5G
Methyl-3-(benzylamino)propanoate
Methyl-3-(benzylamino)propanoate
Vzorec: C₁₁H₁₅NO₂
Bod varu: 145…147 °C/7mmHg
Storage Temperature: Freezer
MDL Number: MFCD01475856
CAS číslo: 23574-01-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR