Tisk…

Methyl-3-[(2-methoxy-2-oxoethyl)amino]propanoate

Dodavatel: Apollo Scientific
Varování

OR300521-10G OR300521-1G OR300521-5G
APOSOR300521-10GEA 0 CZK
APOSOR300521-10G APOSOR300521-1G APOSOR300521-5G
Methyl-3-[(2-methoxy-2-oxoethyl)amino]propanoate
Methyl-3-[(2-methoxy-2-oxoethyl)amino]propanoate
Vzorec: C₇H₁₃NO₄
MW: 175,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11651408
CAS číslo: 105240-68-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR