Tisk…

4-Chloro-5-methyl-isoxazol-3-ylamine

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Amino-4-chloro-5-methylisoxazole

OR300501-1G OR300501-5G
APOSOR300501-1GEA 4730 CZK
APOSOR300501-1G APOSOR300501-5G
4-Chloro-5-methyl-isoxazol-3-ylamine
4-Chloro-5-methyl-isoxazol-3-ylamine
Vzorec: C₄H₅ClN₂O
MW: 132,55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11227222
CAS číslo: 5819-39-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR