Tisk…

5,6,7,8-Tetrahydroquinolin-8-ol

Dodavatel: Apollo Scientific
Varování

OR300368-10G OR300368-1G OR300368-5G
APOSOR300368-10GEA 3580 CZK
APOSOR300368-10G APOSOR300368-1G APOSOR300368-5G
5,6,7,8-Tetrahydroquinolin-8-ol
5,6,7,8-Tetrahydroquinolin-8-ol
Vzorec: C₉H₁₁NO
MW: 149,1897 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369987
CAS číslo: 14631-46-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR