4-Bromoindole-6-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Bromo-1H-indole-6-carboxylic acid
OR300307-1G
OR300307-250MG
APOSOR300307-1GEA
3540
CZK
APOSOR300307-1G
APOSOR300307-250MG
4-Bromoindole-6-carboxylic acid
4-Bromoindole-6-carboxylic acid
Vzorec:
C₉H₆BrNO₂ MW: 240,06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03095205 CAS číslo: 374633-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...