Tisk…

2-Amino-5-bromo-3-methylpyrazine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-Bromo-3-methylpyrazin-2-amine

OR300250-1G OR300250-250MG
APOSOR300250-1GEA 3970 CZK
APOSOR300250-1G APOSOR300250-250MG
2-Amino-5-bromo-3-methylpyrazine
2-Amino-5-bromo-3-methylpyrazine
Vzorec: C₅H₆BrN₃
MW: 188,0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08705773
CAS číslo: 74290-67-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR