Tisk…

5-Formylpicolinonitrile

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Formylpyridine-2-carbonitrile

OR300168-1G OR300168-5G
APOSOR300168-1GEA 2570 CZK
APOSOR300168-1G APOSOR300168-5G
5-Formylpicolinonitrile
5-Formylpicolinonitrile
Vzorec: C7H4N2O
MW: 132.1200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12755972
CAS číslo: 131747-68-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR