3-Hydroxy-5-methylisoxazole 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Hymexazol
OR2998-25G
OR2998-5G
APOSOR2998-25GEA
1780
CZK
APOSOR2998-25G
APOSOR2998-5G
3-Hydroxy-5-methylisoxazole 97%
3-Hydroxy-5-methylisoxazole
Vzorec:
C₄H₅NO₂ MW: 99,09 g/mol Bod tání: 87 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00144468 CAS číslo: 10004-44-1 EINECS: 233-000-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...