Tisk…

4-Methylbenzamidine hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Methylphenylamidine hydrochloride, 4-Methylbenzene-1-carboximidamide hydrochloride, 4-Methylbenzimidamide hydrochloride

OR29962-1G OR29962-5G
APOSOR29962-1GEA 1870 CZK
APOSOR29962-1G APOSOR29962-5G
4-Methylbenzamidine hydrochloride
4-Methylbenzamidine hydrochloride
Vzorec: C₈H₁₁ClN₂
MW: 170.64
Bod tání: 211…214 °C
Storage Temperature: Ambient
MDL Number: MFCD00040183
CAS číslo: 6326-27-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR