6-Chloronicotinohydrazide 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Chloropyridine-5-carbohydrazide
OR29938-1G
APOSOR29938-1GEA
1990
CZK
APOSOR29938-1G
6-Chloronicotinohydrazide 98%
6-Chloronicotinohydrazide
Vzorec:
C₆H₆ClN₃O MW: 171,59 g/mol Bod tání: 250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00221429 CAS číslo: 168893-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...