Tisk…

2,6-Dichloro-N-hydroxybenzenecarboximidoyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: alpha,2,6-Trichlorobenzaldoxime

OR29933-10G OR29933-1G OR29933-5G
APOSOR29933-10GEA 13230 CZK
APOSOR29933-10G APOSOR29933-1G APOSOR29933-5G
2,6-Dichloro-N-hydroxybenzenecarboximidoyl chloride
2,6-Dichloro-N-hydroxybenzenecarboximidoyl chloride
Vzorec: C₇H₄Cl₃NO
MW: 224,48 g/mol
Bod tání: 84…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00673177
CAS číslo: 6579-27-7
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR