Tisk…

4-(4-Chlorophenoxy)aniline

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Amino-4'-chlorodiphenyl ether

OR29829-10G OR29829-50G
APOSOR29829-10GEA 1330 CZK
APOSOR29829-10G APOSOR29829-50G
4-(4-Chlorophenoxy)aniline
4-(4-Chlorophenoxy)aniline
Vzorec: C₁₂H₁₀ClNO
MW: 219,67 g/mol
Bod varu: 205 °C/12mmHg
Bod tání: 99…103 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043925
CAS číslo: 101-79-1
EINECS: 202-976-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR