Ethyl-3-(2-furyl)-3-oxopropanoate
Dodavatel: Apollo Scientific
|
Varování
|
OR29821-1G
OR29821-5G
OR29821-10G
APOSOR29821-1GEA
910
CZK
APOSOR29821-1G
APOSOR29821-5G
APOSOR29821-10G
Ethyl-3-(2-furyl)-3-oxopropanoate
Ethyl-3-(2-furyl)-3-oxopropanoate
Vzorec:
C₉H₁₀O₄ Bod varu: 115…120 °C/4mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00122473 CAS číslo: 615-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...