5-Bromo-2-hydroxy-3-nitropyridine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
5-Bromo-3-nitro-2-pyridinol, 5-Bromo-3-nitropyridin-2(1H)-one
OR29816-100G
APOSOR29816-100GEA
0
CZK
APOSOR29816-100G
5-Bromo-2-hydroxy-3-nitropyridine 97%
5-Bromo-2-hydroxy-3-nitropyridine
Vzorec:
C₅H₃BrN₂O₃ MW: 218,99 g/mol Bod tání: 246…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023473 CAS číslo: 15862-34-7 EINECS: 239-989-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...