6-Methylpyridine-3-carbothioamide
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Methylthionicotinamide
OR29808-1G
OR29808-250MG
APOSOR29808-1GEA
7510
CZK
APOSOR29808-1G
APOSOR29808-250MG
6-Methylpyridine-3-carbothioamide
6-Methylpyridine-3-carbothioamide
Vzorec:
C₇H₈N₂S MW: 152.22 g/mol Bod tání: 193…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD00114709 CAS číslo: 175277-57-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...