Tisk…

Ethyl-2-chloroisonicotinate

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 2-Chloropyridine-4-carboxylate

OR2977-100G
APOSOR2977-100GEA 3870 CZK
APOSOR2977-100G
Ethyl-2-chloroisonicotinate
Ethyl-2-chloroisonicotinate
Vzorec: C₈H₈ClNO₂
MW: 185,6076 g/mol
Bod varu: 130…132 °C/20mmHg
Storage Temperature: Ambient
MDL Number: MFCD04114259
CAS číslo: 54453-93-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR