2-(4-Chlorophenyl)-2-cyanoacetaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(4-chlorophenyl)-3-oxopropanenitrile
OR29596-1G
OR29596-250MG
APOSOR29596-1GEA
3910
CZK
APOSOR29596-1G
APOSOR29596-250MG
2-(4-Chlorophenyl)-2-cyanoacetaldehyde
2-(4-Chlorophenyl)-2-cyanoacetaldehyde
Vzorec:
C₉H₆ClNO MW: 179,61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00052669 CAS číslo: 62538-21-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...