Tisk…

1-(4-Iodophenyl)-1H-pyrrole-2-carbaldehyde

Dodavatel: Apollo Scientific

OR29423-1G OR29423-5G OR29423-10G
APOSOR29423-1GEA 3190 CZK
APOSOR29423-1G APOSOR29423-5G APOSOR29423-10G
1-(4-Iodophenyl)-1H-pyrrole-2-carbaldehyde
1-(4-Iodophenyl)-1H-pyrrole-2-carbaldehyde
Vzorec: C₁₁H₈INO
MW: 297,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01764931
CAS číslo: 261178-18-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR