3-(Chloromethyl)-5-cyclobutyl-1,2,4-oxadiazole
Dodavatel: Apollo Scientific
|
Varování
|
OR29355-250MG
OR29355-1G
OR29355-5G
APOSOR29355-250MGEA
3310
CZK
APOSOR29355-250MG
APOSOR29355-1G
APOSOR29355-5G
3-(Chloromethyl)-5-cyclobutyl-1,2,4-oxadiazole
3-(Chloromethyl)-5-cyclobutyl-1,2,4-oxadiazole
Vzorec:
C₇H₉ClN₂O Storage Temperature: Ambient |
MDL Number:
MFCD00728735 CAS číslo: 259683-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...