Tisk…

Ethyl [(ethoxycarbonyl)thio]acetate

Dodavatel: Apollo Scientific
Varování

OR29339-1G OR29339-2.5G OR29339-5G OR29339-10G
APOSOR29339-1GEA 3740 CZK
APOSOR29339-1G APOSOR29339-2.5G APOSOR29339-5G APOSOR29339-10G
Ethyl [(ethoxycarbonyl)thio]acetate
Ethyl [(ethoxycarbonyl)thio]acetate
Vzorec: C₇H₁₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD00662532
CAS číslo: 52790-15-5
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR