Tisk…

3-(2,4-Dichlorophenyl)propionic acid

Dodavatel: Apollo Scientific

Synonyms: 3-(2,4-Dichlorophenyl)propanoic acid

OR29293-25G
APOSOR29293-25GEA 3640 CZK
APOSOR29293-25G
3-(2,4-Dichlorophenyl)propionic acid
3-(2,4-Dichlorophenyl)propionic acid
Vzorec: C₉H₈Cl₂O₂
MW: 219,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00275605
CAS číslo: 55144-92-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR