2-Hydrazino-3-nitropyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Hydrazinyl-3-nitropyridine
OR29277-1G
OR29277-5G
OR29277-10G
APOSOR29277-1GEA
992
CZK
APOSOR29277-1G
APOSOR29277-5G
APOSOR29277-10G
2-Hydrazino-3-nitropyridine
2-Hydrazino-3-nitropyridine
Vzorec:
C₅H₆N₄O₂ MW: 154,13 g/mol Bod tání: 173…175 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00085923 CAS číslo: 15367-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...