Tisk…

4-Benzoylpiperidine hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: Phenyl(piperidin-4-yl)methanone hydrochloride

OR29257-1G OR29257-5G OR29257-10G
APOSOR29257-1GEA 744 CZK
APOSOR29257-1G APOSOR29257-5G APOSOR29257-10G
4-Benzoylpiperidine hydrochloride
4-Benzoylpiperidine hydrochloride
Vzorec: C₁₂H₁₆ClNO
MW: 225,7200 g/mol
Bod tání: 225…227 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00066982
CAS číslo: 25519-80-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR