Tisk…

Methyl-4-(bromomethyl)benzoate 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Methyl 4-(bromomethyl)benzoate

OR29204-25G OR29204-100G OR29204-5G
APOSOR29204-25GEA 1160 CZK
APOSOR29204-25G APOSOR29204-100G APOSOR29204-5G
Methyl-4-(bromomethyl)benzoate 98%
Methyl-4-(bromomethyl)benzoate
Vzorec: C₉H₉BrO₂
MW: 229,07 g/mol
Bod varu: 130…135 °C/2mmHg
Bod tání: 53…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00032453
CAS číslo: 2417-72-3
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR