Tisk…

Ethyl cyano(hydroxyimino)acetate

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 2-cyano-2-(hydroxyimino)acetate, Ethyl Cyanoglyoxalate Oxime, Cyano(hydroxyimino)acetic Acid Ethyl Ester, Oxyma

OR29185-100G OR29185-250G OR29185-25G
APOSOR29185-100GEA 1240 CZK
APOSOR29185-100G APOSOR29185-250G APOSOR29185-25G
Ethyl cyano(hydroxyimino)acetate
Ethyl cyano(hydroxyimino)acetate
Vzorec: C₅H₆N₂O₃
MW: 142,11 g/mol
Bod tání: 127…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00002112
CAS číslo: 3849-21-6
EINECS: 223-351-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR