5-Chloro-3-chloromethyl[1,2,4]thiadiazole
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR29175-1G
OR29175-5G
APOSOR29175-1GEA
0
CZK
APOSOR29175-1G
APOSOR29175-5G
5-Chloro-3-chloromethyl[1,2,4]thiadiazole
5-Chloro-3-chloromethyl[1,2,4]thiadiazole
Vzorec:
C₃H₂Cl₂N₂S MW: 169,03 g/mol Bod varu: 55 °C/1mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00096470 CAS číslo: 74461-64-6 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...