Tisk…

3-(Chloromethyl)-5-phenyl-1,2,4-oxadiazole

Dodavatel: Apollo Scientific

OR29121-1G OR29121-5G
APOSOR29121-1GEA 2690 CZK
APOSOR29121-1G APOSOR29121-5G
3-(Chloromethyl)-5-phenyl-1,2,4-oxadiazole
3-(Chloromethyl)-5-phenyl-1,2,4-oxadiazole
Vzorec: C₉H₇ClN₂O
Bod tání: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00084968
CAS číslo: 1201-68-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR