Tisk…

5-(2-Pyridyl)-1,3-oxazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-(Pyridin-2-yl)oxazole

OR29044-1G OR29044-250MG
APOSOR29044-1GEA 4960 CZK
APOSOR29044-1G APOSOR29044-250MG
5-(2-Pyridyl)-1,3-oxazole
5-(2-Pyridyl)-1,3-oxazole
Vzorec: C₈H₆N₂O
MW: 146,149 g/mol
Bod varu: 95…98 °C/0,2mmHg
Storage Temperature: Ambient
MDL Number: MFCD00085160
CAS číslo: 70380-73-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR