(4-Methyl-1,3-thiazol-2-yl)acetonitrile
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(4-Methylthiazol-2-yl)acetonitrile
OR28979-10G
OR28979-1G
OR28979-5G
APOSOR28979-10GEA
19680
CZK
APOSOR28979-10G
APOSOR28979-1G
APOSOR28979-5G
(4-Methyl-1,3-thiazol-2-yl)acetonitrile
(4-Methyl-1,3-thiazol-2-yl)acetonitrile
Vzorec:
C₆H₆N₂S MW: 138,19 g/mol Bod varu: 82…83 °C/0,4mmHg Storage Temperature: Ambient |
MDL Number:
MFCD03011392 CAS číslo: 19785-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...