Tisk…

(S)-(+)-Epichlorhydrin

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (S)-(+)-Glycerol epichlorhydrin, (S)-(+)-3-Chloro-1,2-epoxypropane, (S)-(+)-1-Chloro-2,3-epoxypropane, (2S)-2-(chloromethyl)oxirane

OR28971-100G OR28971-25G OR28971-5G
APOSOR28971-100GEA 3810 CZK
APOSOR28971-100G APOSOR28971-25G APOSOR28971-5G
(S)-(+)-Epichlorhydrin
(S)-(+)-Epichlorhydrin
Vzorec: ClCH₂CH(O)CH₂
MW: 92,52 g/mol
Bod varu: 114 °C
Hustota: 1,18 g/cm³ (25 °C)
Bod vzplanutí: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00077760
CAS číslo: 67843-74-7
UN: 2023
ADR: 6.1+3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR