Tisk…

(R)-(-)-Epichlorhydrin 99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (R)-(-)-Glycerol epichlorhydrin, (R)-(-)-3-Chloro-1,2-epoxypropane, (R)-(-)-1-Chloro-2,3-epoxypropane

OR28970-100G OR28970-25G OR28970-500G
APOSOR28970-100GEA 2070 CZK
APOSOR28970-100G APOSOR28970-25G APOSOR28970-500G
(R)-(-)-Epichlorhydrin 99%
(R)-(-)-Epichlorhydrin
Vzorec: ClCH₂CH(O)CH₂
Bod varu: 114…117 °C
Hustota: 1,18 g/cm³ (25 °C)
Bod vzplanutí: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00077759
CAS číslo: 51594-55-9
EINECS: 424-280-2
UN: 2023
ADR: 6.1+3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR