Tisk…

Ethyl-2-chloronicotinate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 2-chloropyridine-3-carboxylate, Ethyl 2-Chloro-3-pyridinecarboxylate, Ethyl 2-chloronicotinate

OR28967-25G OR28967-100G
APOSOR28967-25GEA 622 CZK
APOSOR28967-25G APOSOR28967-100G
Ethyl-2-chloronicotinate 98%
Ethyl 2-chloronicotinate
Vzorec: C₈H₈ClNO₂
MW: 185,61 g/mol
Bod varu: 78 °C/0,08mmHg
Hustota: 1,25 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00674129
CAS číslo: 1452-94-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR