3-Nitrophenyl isocyanate 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR28931-25G
OR28931-5G
APOSOR28931-25GEA
8240
CZK
APOSOR28931-25G
APOSOR28931-5G
3-Nitrophenyl isocyanate 97%
3-Nitrophenyl isocyanate
When melted, may contain small amount of sediment.
Vzorec:
C₇H₄N₂O₃ Bod varu: 130…131 °C/11mmHg Bod tání: 51…52 °C Bod vzplanutí: 113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007220 CAS číslo: 3320-87-4 EINECS: 222-025-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...