N-Benzylethylenediamine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
N1-Benzylethane-1,2-diamine
OR28924-100G
OR28924-25G
APOSOR28924-100GEA
8200
CZK
APOSOR28924-100G
APOSOR28924-25G
N-Benzylethylenediamine 98%
N-Benzylethylenediamine
Vzorec:
C₉H₁₄N₂ Bod varu: 162 °C/20mmHg Hustota: 1 g/cm³ (25 °C) Bod vzplanutí: 95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041896 CAS číslo: 4152-09-4 EINECS: 223-984-5 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...