4-Nitrobenzenesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-Nitrobenzene-1-sulphonyl chloride
OR28919-100G
OR28919-25G
OR28919-5G
APOSOR28919-100GEA
3540
CZK
APOSOR28919-100G
APOSOR28919-25G
APOSOR28919-5G
4-Nitrobenzenesulphonyl chloride
4-Nitrobenzenesulphonyl chloride
Vzorec:
C₆H₄ClNO₄S MW: 221.62 Bod varu: 143…144 °C/1.5mmHg Bod tání: 75…80 °C Hustota: 1.69 Storage Temperature: Ambient |
MDL Number:
MFCD00007445 CAS číslo: 98-74-8 EINECS: 202-697-9 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...