Tisk…

(R)-tert-Butyl 3-((methylsulphonyl)oxy)pyrrolidine-1-carboxylate 95+%

Dodavatel: Apollo Scientific

Synonyms: (R)-3-Methanesulphonyloxy-pyrrolidine-1-carboxylicacid-tert-butyl ester, (R)-3-(Methylsulphonyloxy)pyrrolidine, N-BOC protected

OR2890-10G OR2890-1G OR2890-5G
APOSOR2890-10GEA 12140 CZK
APOSOR2890-10G APOSOR2890-1G APOSOR2890-5G
(R)-tert-Butyl 3-((methylsulphonyl)oxy)pyrrolidine-1-carboxylate 95+%
(R)-tert-Butyl 3-((methylsulphonyl)oxy)pyrrolidine-1-carboxylate
Vzorec: C₁₀H₁₉NO₅S
MW: 265,330 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06809586
CAS číslo: 141699-57-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR