Tisk…

(±)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid 98%

Dodavatel: Apollo Scientific
Varování

OR28891-100G OR28891-25G
APOSOR28891-100GEA 9920 CZK
APOSOR28891-100G APOSOR28891-25G
(±)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid 98%
(±)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Vzorec: C₁₀H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00191496
CAS číslo: 67123-97-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR