Tisk…

(-)-Quinine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol

OR28880-1G OR28880-25G OR28880-5G
APOSOR28880-1GEA 622 CZK
APOSOR28880-1G APOSOR28880-25G APOSOR28880-5G
(-)-Quinine
(-)-Quinine
Vzorec: C₂₀H₂₄N₂O₂
MW: 324.42
Bod tání: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00198096
CAS číslo: 130-95-0
EINECS: 205-003-2
Merck Index: 12,08245

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR