Tisk…

2-(4-Chlorophenoxy)isobutyric acid 97+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(4-Chlorophenoxy)-2-methylpropionic acid, Clofibric acid, 2-(4-Chlorophenoxy)-2-methylpropanoic acid

OR28848-25G OR28848-500G
APOSOR28848-25GEA 3230 CZK
APOSOR28848-25G APOSOR28848-500G
2-(4-Chlorophenoxy)isobutyric acid 97+%
2-(4-Chlorophenoxy)isobutyric acid
Vzorec: C₁₀H₁₁ClO₃
MW: 214,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004192
CAS číslo: 882-09-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR