(R)-(-)-Methyl-3-hydroxybutyrate
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
(R)-Methyl 3-hydroxybutanoate, Methyl (R)-3-hydroxybutyrate, Methyl (R)-(-)-3-Hydroxybutyrate
OR28835-100G
OR28835-10G
OR28835-25G
APOSOR28835-100GEA
7740
CZK
APOSOR28835-100G
APOSOR28835-10G
APOSOR28835-25G
(R)-(-)-Methyl-3-hydroxybutyrate
(R)-(-)-Methyl-3-hydroxybutyrate
Vzorec:
C₅H₁₀O₃ MW: 118,13 g/mol Bod varu: 72 °C/17mmHg Hustota: 1,055 g/cm³ (25 °C) Bod vzplanutí: 73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063289 CAS číslo: 3976-69-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...