Tisk…

(R)-(-)-Glycidyl butyrate

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (-)-[(2R)-Oxiran-2-yl]methyl butanoate, (R)-Oxiran-2-ylmethyl butyrate

OR28822-100G OR28822-25G OR28822-5G
APOSOR28822-100GEA 992 CZK
APOSOR28822-100G APOSOR28822-25G APOSOR28822-5G
(R)-(-)-Glycidyl butyrate
(R)-(-)-Glycidyl butyrate
Vzorec: C₇H₁₂O₃
MW: 144,17 g/mol
Bod varu: 197 °C
Hustota: 1,032 g/cm³ (25 °C)
Bod vzplanutí: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00075120
CAS číslo: 60456-26-0
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR